Learn More
Biomatik Corporation Rat E1A Binding Protein P300 (EP300) ELISA Kit
Supplier:  Biomatik Corporation EKU03820
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Rat. Detection Range: 0.312-20ng/mL. Sensitivity: 0.112ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q91XT0. Target Synonyms: KAT3B; p300; Histone Acetyltransferase p300. Restriction: For Research Use Only.
Catalog No. 50-229-2329