Learn More
Biomatik Corporation Human Elongation Of Very Long Chain Fatty Acids Like Protein 1 (ELOVL1) ELISA Kit
Supplier:  Biomatik Corporation EKU03863
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids.. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.062ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9BW60. Target Synonyms: Ssc1; 3-keto acyl-CoA synthase ELOVL1; ELOVL fatty acid elongase 1; Very-long-chain 3-oxoacyl-CoA synthase 1. Restriction: For Research Use Only.
Catalog No. 50-229-2337