Learn More
Biomatik Corporation Dog Endothelin 1 (EDN1) ELISA Kit
Supplier:  Biomatik Corporation EKU03885
Encompass_Preferred
Dog Endothelin 1 (EDN1) ELISA Kit for detecting EDN1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 6.17-500pg/ml. Sensitivity: 2.84pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P13206. Gene ID: 403424. Target Synonyms: ET1; PPET1; Preproendothelin-1; Big endothelin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2340