Learn More
Biomatik Corporation Guinea pig Eosinophil Chemotactic Factor (ECF) ELISA Kit
Supplier:  Biomatik Corporation EKU03913
Encompass_Preferred
Guinea pig Eosinophil Chemotactic Factor (ECF) ELISA Kit for detecting ECF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Guinea pig (Cavia porcellus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P80325. Gene ID: 100135493. Target Synonyms: CCL11; SCYA11; Eotaxin 1; Eosinophil Chemotactic Protein 1; Chemokine C-C-Motif Ligand 11; Small Inducible Cytokine Subfamily A (Cys-Cys) Member 11. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2344