Learn More
Biomatik Corporation Human Ephrin B1 (EFNB1) ELISA Kit
Supplier:  Biomatik Corporation EKU03924
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.055ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: CFND; CFNS; EFL3; EPLG2; Elk-L; LERK2; EPH-related receptor tyrosine kinase ligand 2; Craniofrontonasal Syndrome(Craniofrontonasal Dysplasia). Restriction: For Research Use Only.
Catalog No. 50-229-2346