Learn More
Biomatik Corporation Mouse Epidermal Growth Factor Receptor (EGFR) ELISA Kit
Supplier:  Biomatik Corporation EKU03937
Encompass_Preferred
Mouse Epidermal Growth Factor Receptor (EGFR) ELISA Kit for detecting EGFR in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q01279. Gene ID: 13649. Target Synonyms: ErbB-1; ErbB1; ER1; ERB-B1; MENA; HER1; Erythroblastic Leukemia Viral (v-Erb-B) oncogene Homolog,Avian; Proto-oncogene c-ErbB-1; Receptor tyrosine-protein kinase erbB-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2348