Learn More
Biomatik Corporation Mouse Receptor Tyrosine Protein Kinase erbB-2 (ErbB2) ELISA Kit
Supplier:  Biomatik Corporation EKU03940
Encompass_Preferred
Mouse Receptor Tyrosine Protein Kinase erbB-2 (ErbB2) ELISA Kit for detecting ErbB2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P70424. Gene ID: 13866. Target Synonyms: CD340; EGFR2; HER-2/neu; HER2; NEU; MLN 19; NGL; TKR1; p185; Epidermal Growth Factor Receptor 2; V-Erb-B2 Erythroblastic Leukemia Viral Oncogene Homolog 2; Neuro/Glioblastoma Derived Oncogene Homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2350