Learn More
Biomatik Corporation Rat Estrogen Receptor Alpha (ERa) ELISA Kit
Supplier:  Biomatik Corporation EKU03975
Encompass_Preferred
Rat Estrogen Receptor Alpha (ERa) ELISA Kit for detecting ERa in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.138ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P06211. Gene ID: 24890. Target Synonyms: ER-A1; ER; ESR; ESR1; ESRA; NR3A1; NR3-A1; Estrogen Receptor 1; Nuclear Receptor Subfamily 3,Group A,Member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2358