Learn More
Biomatik Corporation Human Exportin 5 (XPO5) ELISA Kit
Supplier:  Biomatik Corporation EKU04001
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.121ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9HAV4. Target Synonyms: RANBP21; Exp5; Ran-binding protein 21. Restriction: For Research Use Only.
Catalog No. 50-229-2363