Learn More
Biomatik Corporation Human Fatty Acid Binding Protein 8, Myelin (FABP8) ELISA Kit
Supplier:  Biomatik Corporation EKU04046
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.054ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: PMP2; M-FABP; MFABP; MP2; P2; Peripheral Myelin Protein 2; Myelin P2 Protein. Restriction: For Research Use Only.
Catalog No. 50-229-2367