Learn More
Biomatik Corporation Rat Fibrinogen (FG) ELISA Kit
Supplier:  Biomatik Corporation EKU04092
Encompass_Preferred
Rat Fibrinogen (FG) ELISA Kit for detecting FG in plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 12.5-800ng/ml. Sensitivity: 4.9ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P06399; P14480; P02680. Gene ID: 24366; 24367. Target Synonyms: F1; FI; Fbg; Coagulation Factor I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2374