Learn More
Biomatik Corporation Mouse Fibroblast Growth Factor Receptor 1 (FGFR1) ELISA Kit
Supplier:  Biomatik Corporation EKU04189
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.060ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: CD331; FLT2, KAL2; CEK; FLG; BFGFR, N-SAM,Basic Fibroblast Growth Factor Receptor 1; Fms-Related Tyrosine Kinase-2; Pfeiffer Syndrome. Restriction: For Research Use Only.
Catalog No. 50-229-2384