Learn More
Biomatik Corporation Mouse Fibulin 3 (FBLN3) ELISA Kit
Supplier:  Biomatik Corporation EKU04212
Encompass_Preferred
Sample Type: serum, plasma, urine and other biological fluids. Species Reactivity: Mouse. Detection Range: 15.6-1,000pg/mL. Sensitivity: 6.0pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q8BPB5. Target Synonyms: EFEMP1; DHRD; DRAD; FBNL; MLVT; MTLV; S1-5; EGF-Containing Fibulin-Like Extracellular Matrix Protein 1; Fibrillin-Like; Extracellular protein S1-5. Restriction: For Research Use Only.
Catalog No. 50-229-2388