Learn More
Biomatik Corporation Human Ficolin 2 (FCN2) ELISA Kit
Supplier:  Biomatik Corporation EKU04219
Encompass_Preferred
Human Ficolin 2 (FCN2) ELISA Kit for detecting FCN2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.37ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q15485. Gene ID: 2220. Target Synonyms: EBP37; FCNL; P35; Hucolin; 37 kDa elastin-binding protein; Collagen/Fibrinogen Domain Containing Lectin; Ficolin B; Serum Lectin p35; L-Ficolin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2390