Learn More
Biomatik Corporation Mouse FMS Like Tyrosine Kinase 3 Ligand (Flt3L) ELISA Kit
Supplier:  Biomatik Corporation EKU04241
Encompass_Preferred
Mouse FMS Like Tyrosine Kinase 3 Ligand (Flt3L) ELISA Kit for detecting Flt3L in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P49772. Gene ID: 14256. Target Synonyms: FLT3LG; FLT3-LG; Flt3-L; FL; Fms-Related Tyrosine Kinase 3 ligand; SL cytokine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2394