Learn More
Biomatik Corporation Mouse Forkhead Box Protein A2 (FOXA2) ELISA Kit
Supplier:  Biomatik Corporation EKU04258
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids.. Species Reactivity: Mouse. Detection Range: 0.312-20ng/mL. Sensitivity: 0.126ng/mL.. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P35583. Target Synonyms: FOX-A2; HNF3B; TCF3B; Hepatocyte Nuclear Factor 3,Beta; Transcription factor 3B. Restriction: For Research Use Only.
Catalog No. 50-229-2396