Learn More
Biomatik Corporation Human Four And A Half LIM Domains Protein 1 (FHL1) ELISA Kit
Supplier:  Biomatik Corporation EKU04268
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.058ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: FHL1B; KYO-T; SLIM1; XMPMA; FLH1A; LIM Protein SLIMMER; Skeletal muscle LIM-protein 1. Restriction: For Research Use Only.
Catalog No. 50-229-2399