Learn More
Biomatik Corporation Mouse Free Fatty Acid Receptor 2 (FFAR2) ELISA Kit
Supplier:  Biomatik Corporation EKU04272
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.061ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q8VCK6. Target Synonyms: FFA2R; GPR43; FFA2; GPCR43; G-protein coupled receptor 43. Restriction: For Research Use Only.
Catalog No. 50-229-2401