Learn More
Biomatik Corporation Human G Protein Coupled Estrogen Receptor 1 (GPER) ELISA Kit
Supplier:  Biomatik Corporation EKU04297
Encompass_Preferred
Human G Protein Coupled Estrogen Receptor 1 (GPER) ELISA Kit for detecting GPER in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q99527. Gene ID: 2852. Target Synonyms: GPR30; CMKRL2; DRY12; FEG-1; GPCR-Br; LERGU; LyGPR; Membrane estrogen receptor; Chemoattractant receptor-like 2; Flow-induced endothelial G-protein coupled receptor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2406