Learn More
Biomatik Corporation Human Free Fatty Acid Receptor 4 (FFAR4) ELISA Kit
Supplier:  Biomatik Corporation EKU04300
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids.. Species Reactivity: Human. Detection Range: 0.625-40ng/mL. Sensitivity: 0.252ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: GPR129; GT01; PGR4; GPR120; O3FAR1; G Protein Coupled Receptor 120; Omega-3 fatty acid receptor 1. Restriction: For Research Use Only.
Catalog No. 50-229-2408