Learn More
Biomatik Corporation Human Gastrokine 1 (GKN1) ELISA Kit
Supplier:  Biomatik Corporation EKU04350
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.057ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: A0A8I5KHR1. Target Synonyms: CA11; AMP18; BRICD1; FOV; Foveolin; 18 kDa antrum mucosa protein. Restriction: For Research Use Only.
Catalog No. 50-229-2421