Learn More
Biomatik Corporation Mouse A Disintegrin And Metalloprotease 10 (ADAM10) ELISA Kit
Supplier:  Biomatik Corporation EKU02041
Encompass_Preferred
Mouse A Disintegrin And Metalloprotease 10 (ADAM10) ELISA Kit for detecting ADAM10 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O35598. Gene ID: 11487. Target Synonyms: CD156-C; CD156c; MADM; Kuz; CDw156; Kuzbanian protein homolog; Mammalian disintegrin-metalloprotease. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2045