Learn More
Biomatik Corporation Human Activin AB (ACVAB) ELISA Kit
Supplier:  Biomatik Corporation EKU02115
Encompass_Preferred
Human Activin AB (ACVAB) ELISA Kit for detecting ACVAB in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08476; P09529. Gene ID: 3624; 3625. Target Synonyms: ACV-AB; Activin Beta A Beta B Homodimer. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2055