Learn More
Biomatik Corporation Mouse Adenosine A2b Receptor (ADORA2b) ELISA Kit
Supplier:  Biomatik Corporation EKU02130
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids.. Species Reactivity: Mouse. Detection Range: 78.12-5,000pg/mL. Sensitivity: 30pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-2059