Learn More
Biomatik Corporation Human Glutamate Receptor, Ionotropic, N-Methyl-D-Aspartate 2B (GRIN2B) ELISA Kit
Supplier:  Biomatik Corporation EKU04457
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.117ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q13224. Target Synonyms: NMDAR2B; NR2B; hNR3; Glutamate [NMDA] receptor subunit epsilon-2; N-methyl D-aspartate receptor subtype 2B; N-methyl-D-aspartate receptor subunit 3. Restriction: For Research Use Only.
Catalog No. 50-229-2440