Learn More
Biomatik Corporation Rat Glutathione Peroxidase 3, Plasma (GPX3) ELISA Kit
Supplier:  Biomatik Corporation EKU04481
Encompass_Preferred
Rat Glutathione Peroxidase 3, Plasma (GPX3) ELISA Kit for detecting GPX3 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.37ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P23764. Gene ID: 64317. Target Synonyms: GSHPX3; GPx-P; GSHPx-P; GPXP; Plasma glutathione peroxidase; Extracellular glutathione peroxidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2443