Learn More
Biomatik Corporation Human Glutathione S Transferase Theta 1 (GSTt1) ELISA Kit
Supplier:  Biomatik Corporation EKU04499
Encompass_Preferred
Human Glutathione S Transferase Theta 1 (GSTt1) ELISA Kit for detecting GSTt1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.064ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P30711. Gene ID: 2952. Target Synonyms: GST-T1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2448