Learn More
Biomatik Corporation Human Glycine Transporter 1 (GLYT1) ELISA Kit
Supplier:  Biomatik Corporation EKU04512
Encompass_Preferred
Sample Type: Tissue homogenates and other biological fluids.. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.134ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: SLC6A9; Solute Carrier Family 6 Member 9,Neurotransmitter Transporter,Glycine; Sodium And Chloride Dependent Glycine Transporter 1. Restriction: For Research Use Only.
Catalog No. 50-229-2450