Learn More
Biomatik Corporation Mouse Glycogen Phosphorylase, Liver (PYGL) ELISA Kit
Supplier:  Biomatik Corporation EKU04516
Encompass_Preferred
Mouse Glycogen Phosphorylase, Liver (PYGL) ELISA Kit for detecting PYGL in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9ET01. Gene ID: 110095. Target Synonyms: GPLL; GPBB; Hers Disease; Glycogen Storage Disease Type VI. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2452