Learn More
Biomatik Corporation Human Glypican 1 (GPC1) ELISA Kit
Supplier:  Biomatik Corporation EKU04541
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 0.78-50ng/mL. Sensitivity: 0.29ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P35052. Target Synonyms: Glypican Proteoglycan 1; Secreted glypican-1. Restriction: For Research Use Only.
Catalog No. 50-229-2455