Learn More
Biomatik Corporation Human Granzyme M (GZMM) ELISA Kit
Supplier:  Biomatik Corporation EKU04570
Encompass_Preferred
Human Granzyme M (GZMM) ELISA Kit for detecting GZMM in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P51124. Gene ID: 3004. Target Synonyms: GZM-M; LMET1; MET1; Hu-Met-1; Met-ase; Lymphocyte Met Ase 1; Met-1 serine protease; Natural killer cell granular protease. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2463