Learn More
Biomatik Corporation Rat Heat Shock 70kDa Protein 1A (HSPA1A) ELISA Kit
Supplier:  Biomatik Corporation EKU04657
Encompass_Preferred
Rat Heat Shock 70kDa Protein 1A (HSPA1A) ELISA Kit for detecting HSPA1A in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.55ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P0DMW0. Gene ID: 24472. Target Synonyms: HSP70-1; HSP-A1A; HSPA1; HSP70-2; HSX70. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2476