Learn More
Biomatik Corporation Human Heat Shock Protein Beta 6 (HSPb6) ELISA Kit
Supplier:  Biomatik Corporation EKU04688
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 0.781-50ng/mL. Sensitivity: 0.29ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: O14558. Target Synonyms: HSP20; Heat Shock Protein 20 kD; Heat Shock Protein B6, Alpha Crystallin Related; Heat shock 20 kDa-like protein p20. Restriction: For Research Use Only.
Catalog No. 50-229-2479