Learn More
Biomatik Corporation Rat Heparin Cofactor II (HCII) ELISA Kit
Supplier:  Biomatik Corporation EKU04745
Encompass_Preferred
Rat Heparin Cofactor II (HCII) ELISA Kit for detecting HCII in plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.153ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q64268. Gene ID: 79224. Target Synonyms: SERPIND1; HC2; HCF2; HC II; HLS2; LS2; Serpin D1; Serpin Peptidase Inhibitor Clade D Member 1; Protease inhibitor leuserpin-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2487