Learn More
Biomatik Corporation Mouse Histone Deacetylase 4 (HDAC4) ELISA Kit
Supplier:  Biomatik Corporation EKU04817
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.312-20ng/mL. Sensitivity: 0.113ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q6NZM9. Target Synonyms: HD4; HDAC-A; HDACA. Restriction: For Research Use Only.
Catalog No. 50-229-2498