Learn More
Biomatik Corporation Horse Immunoglobulin G (IgG) ELISA Kit
Supplier:  Biomatik Corporation EKU04892
Encompass_Preferred
Horse Immunoglobulin G (IgG) ELISA Kit for detecting IgG in serum, plasma, other biological fluids. Species Reactivity: Horse (Equus caballus; Equine). Detection Range: 6.17-500ug/ml. Sensitivity: 2.99ug/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2508