Learn More
Biomatik Corporation Pig Insulin Like Growth Factor Binding Protein 3 (IGFBP3) ELISA Kit
Supplier:  Biomatik Corporation EKU04996
Encompass_Preferred
Pig Insulin Like Growth Factor Binding Protein 3 (IGFBP3) ELISA Kit for detecting IGFBP3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 6.25-400ng/ml. Sensitivity: 2.52ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P16611. Gene ID: 448812. Target Synonyms: BP53; IBP3; Growth Hormone-Dependent Binding Protein; Acid Stable Subunit Of The 140 K IGF Complex. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2525