Learn More
Biomatik Corporation Human Integrin Alpha 2B (ITGa2B) ELISA Kit
Supplier:  Biomatik Corporation EKU05031
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.060ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P08514. Target Synonyms: CD41; CD41B; ITG-A2B; GTA; GP2B; GPIIb; HPA3; Platelet Glycoprotein IIb Of IIb/IIIa Complex; Antigen CD41. Restriction: For Research Use Only.
Catalog No. 50-229-2528