Learn More
Biomatik Corporation Human Interferon Alpha/Beta Receptor 2 (IFNa/bR2) ELISA Kit
Supplier:  Biomatik Corporation EKU05086
Encompass_Preferred
Sample Type: Serum, plasma, urine and other biological fluids.. Species Reactivity: Human. Detection Range: 0.625-40ng/mL. Sensitivity: 0.262ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P48551. Target Synonyms: IFNAR2; IFN-alpha-REC; IFN-R; IFNABR; IFNARB; Interferon(Alpha,Beta And Omega)Receptor Beta Chain; Interferon alpha binding protein; Type I interferon receptor 2. Restriction: For Research Use Only.
Catalog No. 50-229-2537