Learn More
Biomatik Corporation Pig Interleukin 1 Alpha (IL1a) ELISA Kit
Supplier:  Biomatik Corporation EKU05132
Encompass_Preferred
Pig Interleukin 1 Alpha (IL1a) ELISA Kit for detecting IL1a in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P18430. Gene ID: 397094. Target Synonyms: IL1-A; IL-1α; IL1F1; Preinterleukin 1 Alpha; Hematopoietin-1; Pro-Interleukin-1-Alpha; Interleukin-1 Family Member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2544