Learn More
Biomatik Corporation Bovine Interleukin 13 (IL13) ELISA Kit
Supplier:  Biomatik Corporation EKU05215
Encompass_Preferred
Bovine Interleukin 13 (IL13) ELISA Kit for detecting IL13 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9XSV9. Gene ID: 281247. Target Synonyms: ALRH; BHR1; P600; IL-13; NC30. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2556