Learn More
Biomatik Corporation Dog Interleukin 3 (IL3) ELISA Kit
Supplier:  Biomatik Corporation EKU05313
Encompass_Preferred
Dog Interleukin 3 (IL3) ELISA Kit for detecting IL3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9BDX4. Gene ID: 481497. Target Synonyms: MCGF; MULTI-CSF; MCSF; M-CSF; Multiple; Multilineage-Colony-Stimulating Factor; Hematopoietic Growth Factor; P-Cell Stimulating Factor; Mast-Cell Growth Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2568