Learn More
Biomatik Corporation Mouse Involucrin (INV) ELISA Kit
Supplier:  Biomatik Corporation EKU05395
Encompass_Preferred
Mouse Involucrin (INV) ELISA Kit for detecting INV in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P48997. Target Synonyms: IVL. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2580