Learn More
Biomatik Corporation Rat Janus Kinase 2 (JAK2) ELISA Kit
Supplier:  Biomatik Corporation EKU05415
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Rat. Detection Range: 0.156-10ng/mL. Sensitivity: 0.059ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q62689. Target Synonyms: JTK10; Tyrosine-protein kinase JAK2. Restriction: For Research Use Only.
Catalog No. 50-229-2586