Learn More
Biomatik Corporation Human Cytokeratin 20 (CK 20) ELISA Kit
Supplier:  Biomatik Corporation EKU05473
Encompass_Preferred
Human Cytokeratin 20 (CK 20) ELISA Kit for detecting CK 20 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P35900. Gene ID: 54474. Target Synonyms: KRT20; Keratin 20; Protein IT; Keratin, type I cytoskeletal 20. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2593