Learn More
Biomatik Corporation Human Keratin 6A (KRT6A) ELISA Kit
Supplier:  Biomatik Corporation EKU05478
Encompass_Preferred
Human Keratin 6A (KRT6A) ELISA Kit for detecting KRT6A in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P02538. Gene ID: 3853. Target Synonyms: K6A; KRT6C; KRT6D; CK6C; K6C; CK6D; K6D; Keratin 6C; Keratin 6D; Keratin, type II cytoskeletal 6A; Type-II keratin Kb6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2595