Learn More
Biomatik Corporation Rat Kininogen 1 (KNG1) ELISA Kit
Supplier:  Biomatik Corporation EKU05503
Encompass_Preferred
Rat Kininogen 1 (KNG1) ELISA Kit for detecting KNG1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08934. Target Synonyms: BDK; KNG; HMWK; Alpha-2-Thiol Proteinase Inhibitor; Bradykinin; Kallidin; Fitzgerald factor; High molecular weight kininogen; Williams-Fitzgerald-Flaujeac factor; Ile-Ser-Bradykinin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2598