Learn More
Biomatik Corporation Human Kynurenine-3-Monooxygenase (KMO) ELISA Kit
Supplier:  Biomatik Corporation EKU05522
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.78-50ng/mL. Sensitivity: 0.30ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: O15229. Target Synonyms: Kynurenine 3-Hydroxylase. Restriction: For Research Use Only.
Catalog No. 50-229-2600