Learn More
Biomatik Corporation Human Lactamase Beta (LACTb) ELISA Kit
Supplier:  Biomatik Corporation EKU05527
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.114ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P83111. Target Synonyms: G24; MRPL56; Mitochondrial Ribosomal Protein L56; Serine beta-lactamase-like protein LACTB, mitochondrial. Restriction: For Research Use Only.
Catalog No. 50-229-2602