Learn More
Biomatik Corporation Rat Lectin Like Oxidized Low Density Lipoprotein Receptor 1 (LOX1) ELISA Kit
Supplier:  Biomatik Corporation EKU05578
Encompass_Preferred
Rat Lectin Like Oxidized Low Density Lipoprotein Receptor 1 (LOX1) ELISA Kit for detecting LOX1 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 62.5-4,000pg/ml. Sensitivity: 22.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O70156. Gene ID: 140914. Target Synonyms: OLR1; CLEC8A; SCARE1; Oxidized low-density lipoprotein receptor 1; C-type lectin domain family 8 member A; Lectin-like oxidized LDL receptor 1; Lectin-type oxidized LDL receptor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2609